Learn More
Electron Microscopy Sciences Grid Boxes for use in routine grid handling and long term storage. Capable of storing 3.05 mm or 2.3 mm diameter grids, 25 column card.
SDP
Supplier:  Electron Microscopy Sciences 7115910
Encompass_Preferred
Our grid boxes are for use in routine grid handling and long term storage. Each diamond will hold Stores 3.05 mm or 2.3 mm diameter grids. The two side features allow easier handling during specimen removal. Grid can be catalogued using the alpha numeric index on the side and the durable 25 column card for record keeping. EMS25 is a small grid storage box, designed as a low alternative to larger grid boxes.
Catalog No. NC3815455