Learn More
Medchemexpress LLC EMT inhibitor-2 | 2232228-60-1 | 98.8% | 1 ML

Supplier: Medchemexpress LLC HY12885910MM/1ML
EMT inhibitor-2, also known as Compound 1, is an agent that inhibits epithelial-mesenchymal transition (EMT) induced by immunocyte-released substances like IL-1β and TGF-β. This compound also demonstrates inhibitory effects on CYP3A4 (testosterone) and CYP2C9.
- Inhibits epithelial-mesenchymal transition
- Inhibits CYP3A4 and CYP2C9 enzymes
- Molecular weight of 470.47
- Chemical formula: C24H26N2O8
- Appearance: Off-white to light yellow solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.