Learn More
MSC Emuge Plug Thread Gage, M2.5x0.45 Thread, 6H Class, Double End, Go/No-Go
Supplier:  MSC 29090966
Encompass
This plug thread gage is designed for precise measurement with an M2.5x0.45 thread and 6H class. It features a double-end design, incorporating both "Go" and "No Go" gaging elements to ensure accurate thread inspection. A handle is included for ease of use.
  • Go and no go gaging
  • Thread size M2.5x0.45
  • Made from tool steel
  • 6H class for precision
  • Handle included for convenience
  • Conforms to DIN ISO 1502 standards
  • Right hand threading
  • Bright/uncoated finish
  • Double end design
  • Includes short form certification
  • Manufactured in Germany
Catalog No. 50-005-29528