Learn More
MSC Emuge Plug Thread Gage, M30x1.5, 6H, Double End, Go/No Go
Supplier: MSC 15985179
The Emuge Plug Thread Gage is designed for M30x1.5 thread, featuring a 6H class. This double-ended gage functions as both 'Go' and 'No Go' for inspection and includes a handle. Made of tool steel, it conforms to DIN ISO 1502 standards, has right-hand threading, and a bright/uncoated finish. It also includes a short form certification and is manufactured in Germany.
- Functions as both Go and No Go
- M30x1.5 thread size
- Constructed from tool steel
- 6H classification
- Includes a handle
- Conforms to DIN ISO 1502 standards
- Right-hand threading
- Bright/uncoated finish
- Double-ended design
- Includes short form certification