Learn More
Medchemexpress LLC Emvododstat | 1256565-36-2 | 99.9% | 467.34 g·mol-1 | C25H20Cl2N2O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY12459310MG
Encompass_Preferred
PTC299 (Emvododstat) is a small-molecule research inhibitor that suppresses vascular endothelial growth factor (VEGF) protein translation and inhibits dihydroorotate dehydrogenase (DHODH). Supplied as a 10 mg powder with high reported purity, it is intended for laboratory research use and includes specified storage recommendations. CAS 1256565-36-2; molecular formula C25H20Cl2N2O3; molecular weight 467.34 g·mol-1.
  • Inhibits VEGF protein translation and DHODH activity.
  • High reported purity suitable for research use.
  • Provided as a 10 mg powder for laboratory applications.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Includes CAS identifier for unambiguous compound tracking.
Catalog No. 50-002-36205