Learn More
Medchemexpress LLC En219-alkyne | 99.6% | 488.76 g·mol⁻¹ | C22H19BrClN3O3 | 5 MG

Supplier: Medchemexpress LLC HY115715A5MG
EN219-alkyne is an alkyne-functionalized covalent probe designed to engage RNF114 and enable click-chemistry labeling and activity-based protein profiling. Supplied as a light brown solid with high purity, it provides a reactive alkyne handle for biochemical conjugation and target identification studies.
- High purity of 99.62%.
- Alkyne handle for click-chemistry conjugation.
- Targets RNF114 as a covalent ligand for activity profiling.
- Molecular formula C22H19BrClN3O3 and molecular weight 488.76 g·mol⁻¹.
- Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
- Store sealed at -20°C; in solution store at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.