Learn More
TARGETMOL CHEMICALS INC EN3356 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T301615MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. EN3356 is an orally available and selective inhibitor of steroidal 17-alpha-hydroxylase/C1720 cleavage enzyme (CYP17A1 or CYP17) a non-steroidal cleavage enzyme-selective compound with potential anti-androgenic and anti-tumor activity. purity: 97%
Catalog No. 50-258-0428