Learn More
Medchemexpress LLC En450 Nf-κb degrader | 793719-01-4 | 98.6% | C11H13ClN2O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY494445MG
Encompass_Preferred
EN450 is a cysteine-reactive covalent molecular glue degrader that targets NF-κB. It interacts with the allosteric C111 in the E2 ubiquitin ligase UBE2D and induces the formation of a ternary complex between UBE2D and NFKB1. This compound achieves its anti-proliferative effects through a Cullin E3 ligase and proteasome-dependent mechanism, significantly inhibiting the proliferation of HAP1 leukemia cancer cells and HEK293T cells, and reducing the levels of the p105 and p50 subunits of NFκB1.
  • Targets Nf-κb
  • Interacts with allosteric C111 in the E2 ubiquitin ligase UBE2D
  • Induces ternary complex formation between UBE2D and NFKB1
  • Achieves anti-proliferative effects via Cullin E3 ligase and proteasome-dependent mechanism
  • Inhibits proliferation of HAP1 leukemia cancer cells and HEK293T cells
  • Reduces levels of p105 and p50 subunits of NFκB1
Catalog No. 50-003-57794