Learn More
Medchemexpress LLC HY-B0331A 500mg Medchemexpress, Enalapril (maleate) CAS:76095-16-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0331A500MG
Encompass_Preferred
Medchemexpress, HY-B0331A 500mg Enalapril (maleate) CAS:76095-16-4 Enalapril (maleate) (MK-421 (maleate)), the active metabolite of enalapril, is an angiotensin-converting enzyme (ACE) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2475