Learn More
AdipoGen Enalaprilat
SDP
Supplier:  AdipoGen CDXE0090M050
Encompass_Preferred
Chemical. CAS 76420-72-9. Formula C18H24N2O5. MW 348.4. Synthetic. Active metabolite enalapril. Nonsulfhydryl angiotensin-converting enzyme ACE inhibitor. Shown to improve cardiac preservation via bradykinin receptor and PKC without affecting ATP metabolism. Has some antioxidant activity, which does not include the PKC-NADPH oxidase pathway. Increases PPARbeta/delta expression, without influence on PPARalpha and PPARgamma.
Catalog No. 50-256-0970