Learn More
Selleck Chemical LLC Enalaprilat Dihydrate S1657-1g
SDP
Supplier:  Selleck Chemical LLC S16571G
Encompass_Preferred
Enalaprilat (MK-422) is an angiotensin-converting enzyme (ACE) inhibitor with IC50 of 1 94 nM
Catalog No. 50-311-0954