Learn More
TARGETMOL CHEMICALS INC ENALAPRILAT DIHYDRATE 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6340200MG
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Enalaprilat Dihydrate (MK-422 Dihydrate) (IC50=1.94 nM) is a potent angiotensin-converting enzyme (ACE) inhibitor. purity: 99%
Catalog No. 50-257-9424