Learn More
Selleck Chemical LLC Enalaprilat Dihydrate 50mg 84680-54-6 MK-422 Dihydrate
SDP
Supplier:  Selleck Chemical LLC S165750MG
Encompass_Preferred
Enalaprilat (MK-422) is an angiotensin-converting enzyme (ACE) inhibitor with IC50 of 1.94 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1492