Learn More
Apexbio Technology LLC Enalaprilat Dihydrate 84680-54-6 50mg
SDP
Supplier:  Apexbio Technology LLC B220450
Encompass_Preferred
Enalaprilat Dihydrate is a small-molecule inhibitor targeting angiotensin-converting enzyme (ACE) It is designed to inhibit ACE thereby blocking the conversion of angiotensin I to the potent vasoconstrictor angiotensin II and reducing vascular resistance and blood pressure Enalaprilat Dihydrate exerts its biological activity primarily through competitive binding to the active site of ACE It demonstrates high potency and specificity with an inhibitory concentration (IC50) value in the low nanomolar range Based on these pharmacological properties Enalaprilat Dihydrate holds research potential in studies of ACE inhibition mechanisms cardiovascular physiology and drug discovery targeting hypertension-related pathologies
Catalog No. 50-000-91010