Learn More
Medchemexpress LLC Enalaprilat dihydrate | 84680-54-6 | 98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0231A
Encompass_Preferred
Enalaprilat dihydrate (MK-422) is the active metabolite of the oral proagent Enalapril. It is a potent, competitive, and long-acting angiotensin-converting enzyme (ACE) inhibitor, making it suitable for hypertension research.
  • Potent, competitive, and long-acting ACE inhibitor
  • IC50 of 1.94 nM for ACE
  • Attenuates IGF-I induced neonatal rat cardiac fibroblast growth in vitro
  • Shows significant intraocular pressure (IOP)-lowering effect in rabbits in vivo
Catalog No. 50-004-50602