Learn More
Medchemexpress LLC Enalaprilat (dihydrate) | 84680-54-6 | 99.6% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB023110MM/1ML
Encompass_Preferred
Enalaprilat dihydrate (MK-422), the active metabolite of the oral proagent Enalapril, is a potent, competitive, and long-acting angiotensin-converting enzyme (ACE) inhibitor with an IC50 of 1.94 nM. This product is intended for research on hypertension.
  • Potent and long-acting ACE inhibitor
  • Attenuates IGF-I induced neonatal rat cardiac fibroblast growth in vitro
  • Shows significant intraocular pressure (IOP)-lowering effect in rabbits in vivo
Catalog No. 50-004-05648