Learn More
Medchemexpress LLC Enalaprilat dihydrate | 84680-54-6 | 99.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0231100MG
Encompass_Preferred
Enalaprilat dihydrate (MK-422) is the active metabolite of the oral proagent Enalapril. It functions as a potent, competitive, and long-acting angiotensin-converting enzyme (ACE) inhibitor with an IC50 of 1.94 nM. This product can be used for the research of hypertension.
  • Potent, competitive, and long-acting ACE inhibitor
  • Attenuates IGF-I induced neonatal rat cardiac fibroblast growth
  • Shows significant intraocular pressure (IOP)-lowering effect in rabbits
  • Utilized in research related to hypertension
Catalog No. 50-004-05778