Learn More
Medchemexpress LLC Enalaprilat dihydrate | 84680-54-6 | 99.6% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0231500MG
Encompass_Preferred
Enalaprilat dihydrate, the active metabolite of Enalapril, is a potent, competitive, and long-acting angiotensin-converting enzyme (ACE) inhibitor with an IC50 of 1.94 nM. It is used for hypertension research.
  • Potent, competitive, and long-acting ACE inhibitor
  • Attenuates IGF-I induced neonatal rat cardiac fibroblast growth
  • Demonstrates a significant intraocular pressure (IOP)-lowering effect
Catalog No. 50-004-06427