Learn More
Medchemexpress LLC Enalaprilat (MK-422 anhydrous) | 76420-72-9 | 99.76% | 100 MG

Supplier: Medchemexpress LLC HYB0231A100MG
Enalaprilat (MK-422 anhydrous) is the active metabolite of the oral proagent Enalapril. It is a potent, competitive, and long-acting angiotensin-converting enzyme (ACE) inhibitor with an IC50 of 1.94 nM. This product is for research use only and can be used for the research of hypertension.
- Molecular weight: 348.39
- Formula: C18H24N2O5
- Appearance: Solid
- Color: White to off-white
- IC50: 1.94 nM (ACE)
- Attenuates IGF-I induced neonatal rat cardiac fibroblast growth in a concentration-dependent manner
- Demonstrates a significant intraocular pressure (IOP)-lowering effect in rabbits
- Powder storage: -20°C for 3 years, 4°C for 2 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
- Shipping: Room temperature in continental US
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.