Learn More
Cayman Chemical EnalaprIlathydrate 10mg
SDP
Supplier:  Cayman Chemical 1760310 MG
Encompass_Preferred
The active metabolite of the ACE inhibitor, enalapril; inhibits ACE activity with an IC50 value of 5.8 nM in vitro
Catalog No. 50-296-8366