Learn More
Selleck Chemical LLC Enarodustat S9656-5MG
SDP
Supplier:  Selleck Chemical LLC S96565MG
Encompass_Preferred
Enarodustat (JTZ-951) is a potent and orally active HIF prolyl hydroxylase inhibitor with IC50 of 0 22 M for PHD2 and EC50 of 5 7 M for EPO release from Hep3B cells Enarodustat has the potential for the treatment of renal anemia
Catalog No. 50-000-23953