Learn More
Selleck Chemical LLC Enasidenib
SDP
Supplier:  Selleck Chemical LLC S820510MM/1ML
Encompass_Preferred
Enasidenib (AG-221) is a first-in-class oral potent reversible selective inhibitor of the IDH2 mutant enzyme
Catalog No. 50-000-01910