Learn More
Medchemexpress LLC Enasidenib mesylate | 1650550-25-6 | MFCD30489472 | 99.9% | 569.48 g/mol | C20H21F6N7O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY18690A10MG
Encompass_Preferred
Enasidenib mesylate is a reversible, selective inhibitor of mutant isocitrate dehydrogenase 2 (IDH2) used in research to study IDH2-mutant leukemia biology and differentiation. It reverses mutant IDH2-mediated epigenetic changes and impairs self-renewal of mutant leukemia cells. Supplied as a high-purity solid for in vitro applications.
  • Potent, reversible inhibitor of mutant IDH2.
  • Reverses mutant IDH2 effects on DNA methylation.
  • High purity suitable for biochemical and cellular assays.
  • Solid form for stable storage and handling.
  • Recommended solvent: DMSO for sample preparation.
  • Store sealed, away from moisture to maintain stability.
Catalog No. 50-002-94997