Learn More
TARGETMOL CHEMICALS INC ENBEZOTINIB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T622855MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Enbezotinib is an inhibitor of RET autophosphorylation and can be used in cancer research. purity: 99%
Catalog No. 50-257-9478