Learn More
Medchemexpress LLC HY-107346 100mg Medchemexpress, Enbucrilate CAS:6606-65-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107346100MG
Encompass_Preferred
Medchemexpress, HY-107346 100mg Enbucrilate CAS:6606-65-1 Enbucrilate (Butyl cyanoacrylate) is a cyanoacrylate ester that has been used as surgical tissue adhesive. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2280