Learn More
Medchemexpress LLC Encaleret | 787583-71-5 | 99.9% | C29H33ClFNO4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1440110MM/1ML
Encompass_Preferred
Encaleret is an orally active antagonist of the calcium-sensing receptor (CaSR) with an IC₅₀ of 12 nM. It functions by inhibiting the excessive activity of functional gain-of-function CaSR variants. This compound is primarily used in research for conditions such as osteoporosis and autosomal dominant hypocalcemia type 1.
  • Restores blood calcium levels
  • Promotes parathyroid hormone secretion
  • Improves magnesium and phosphorus metabolism
  • Increases urinary calcium excretion
Catalog No. 50-003-56163