Learn More
Hach Company Encap PPS pH Sens-LCP-5-Wire
Supplier:  Hach Company 6028P010F00000N
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Gel Filled Standard-10 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode Gel Filled Standard
Catalog No. 50-002-07858