Learn More
ATCC Encephalomyocarditis virus (M-variant) - cell culture-passaged viral stock
Supplier: ATCC VR1479
Encephalomyocarditis virus (M-variant) is a tissue culture-passaged viral isolate used as a murine infection model and for studies of virus-induced diabetes. Supplied as a frozen stock for propagation in susceptible cell lines under standard tissue culture conditions.
- M-variant, tissue culture passaged.
- Propagate on NCTC clone 929 (L-929) cells.
- Complete medium: EMEM with 10% fetal bovine serum.
- Incubation: 4-10 days at 37°C in 5% CO2.
- Storage: -70°C or colder.
- May lose diabetogenic properties after multiple cell culture passages.
- Source: originally isolated from mouse heart.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.