Learn More
Medchemexpress LLC HY-13646 10mg Medchemexpress, Encequidar CAS:849675-66-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1364610MG
Encompass_Preferred
Medchemexpress, HY-13646 10mg Encequidar CAS:849675-66-7 Encequidar (HM30181; HM30181A) is a potent and selective inhibitor of P-glycoprotein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9017