Learn More
Medchemexpress LLC Encequidar mesylate | 849675-87-2 | MFCD25976626 | 99.8% | C39H40N6O10S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13646A10MG
Encompass_Preferred
Encequidar mesylate is a competitive and selective P-glycoprotein (P-gp) inhibitor used in research to investigate P-gp-mediated drug transport and to evaluate strategies for improving oral absorption of P-gp substrate drugs. Supplied as the mesylate salt with confirmed high purity for analytical and preclinical studies.
  • Competitive and selective P-glycoprotein inhibitor.
  • Mesylate salt form for handling and stability.
  • Facilitates studies of intestinal and cellular P-gp transport.
  • Used to evaluate oral bioavailability enhancement strategies.
  • High purity (99.8%) for consistent experimental results.
  • Chemical formula C39H40N6O10S and molecular weight 784.83 g/mol.
  • Suitable for in vitro and in vivo preclinical research.
Catalog No. 50-002-59219