Learn More
TARGETMOL CHEMICALS INC Enclomiphene citrate 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T731325MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Enclomiphene citrate (trans-Clomiphene citrate) is a non-steroidal estrogen receptor antagonist,with antioestrogenic property Purity 99.95%
Catalog No. 50-288-0143