Learn More
Medchemexpress LLC Enclomiphene-d4 | 15690-57-0 | 99.9% | 409.98 | 1MG
SDP
Supplier:  Medchemexpress LLC HY118861S11MG
Encompass_Preferred
Enclomiphene-d4 is a deuterium labeled Enclomiphene. Enclomiphene is a potent and orally active estrogen receptor antagonist with antioestrogenic property. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Enclomiphene
  • Potent and orally active estrogen receptor antagonist with antioestrogenic property
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-37113