Learn More
Grainger End Mill Kyocera EM 12503F 1.2500 Dia
Supplier:  Grainger 170HC7
Encompass_Preferred
The Kyocera 170HC7 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, contouring, and face milling in both ferrous and non-ferrous materials. The end mill features a center cutting design for plunge cuts.
Catalog No. 50-276-3680