Learn More
Grainger End Mill Kyocera EM 15002000LE 1.5000
Supplier:  Grainger 170HD1
Encompass_Preferred
The Kyocera 170HD1 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, and contouring in steels and cast irons. The end mill features a center cutting design for plunge cuts and slotting.
Catalog No. 50-276-4753