Learn More
Grainger End Mill Kyocera MEC 1000S10011T 10
Supplier:  Grainger 170HF7
Encompass_Preferred
The Kyocera 170HF7 is a 1/2" diameter, 4 flute, high feed end mill made of micrograin carbide with TiAIN coating. It has a 1-1/2" LOC and can be run at speeds up to 500 SFM. The end mill features variable helix flutes to reduce harmonics and chatter and is designed for high feed milling in steel, stainless steel, and cast iron. It has a tolerance of +/- .002" on diameter and is made in Japan by Kyocera. The end mill comes with a 30 day warranty against manufacturer defects.
Catalog No. 50-276-4833