Learn More
Grainger End Mill Kyocera MEC 1500S12517 1.5000
Supplier:  Grainger 170HH4
Encompass_Preferred
The Kyocera 170HH2 is a 2 flute, high helix end mill made of micrograin carbide with TiAlN coating. It has a 1/2" shank diameter, 2" overall length, and 1" cutting diameter. The end mill features variable helix flutes to reduce harmonics and vibration and is designed for high feed milling in steel, stainless steel, and cast iron. With its high helix geometry, this end mill is well-suited for efficient slotting, profiling, and shoulder milling applications.
Catalog No. 50-276-4677