Learn More
Grainger End Mill Kyocera MECX 10S10071T 10m
Supplier:  Grainger 170HP0
Encompass_Preferred
The Kyocera 170HN0 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, and contouring in steels, cast iron, aluminum, and other non-ferrous metals. The end mill features a center cutting design for plunging.
Catalog No. 50-276-4354