Learn More
Grainger End Mill Kyocera MEW 1000W100103T 1
Supplier:  Grainger 170HJ4
Encompass_Preferred
The KYOCERA Indexable Face Mill 80.00mm is a high-performance face milling cutter with a diameter of 80mm. It features indexable carbide inserts with 8 cutting edges for long tool life. The cutter body is made of alloy steel and balanced for smooth milling. It has a right-hand spiral design with through coolant capability. The arbor hole diameter is 32mm. Recommended cutting parameters are 12,000 RPM max with 0.2mm/tooth feed rate. This durable and precise face mill is well-suited for heavy roughing and finishing of steel, stainless steel, cast iron, and other hard materials.
Catalog No. 50-276-3684