Learn More
Grainger End Mill Kyocera MFWN 902000RW1253T 2.
Supplier:  Grainger 170HH7
Encompass_Preferred
The Kyocera Indexable Face Mill is a 2 inch diameter face milling cutter with 4 indexable carbide inserts. It has a 1 inch shank diameter and comes with a Weldon flat and keyway for secure tool holding. The cutter body is made of steel and can mill a variety of ferrous and non-ferrous materials. With the indexable carbide inserts, it provides efficient and economical face milling by allowing quick insert changes as they wear. This durable Kyocera face mill is well-suited for heavy duty milling applications.
Catalog No. 50-276-4279