Learn More
ATCC End1/E6E7; Endocervix; Human (Homo sapiens)
Supplier:  ATCC CRL2615
Encompass
End1/E6E7 is an epithelial cell line that was isolated in 1996 from the endocervix of a 43-year-old, female patient undergoing a hysterectomy for endometriosis. This cell line was deposited by D Anderson, RN Fichorova, and JG Rheinwald.
Catalog No. 50-238-1894
 Restricted Item