Learn More
Tosoh Bioscience LLC ENDFITTING 10UM FRIT 7.5MMID
Supplier:  Tosoh Bioscience LLC 0005748
Encompass
Fitting, End; With fixed 10 micrometer frit for 7.5mm I.D. columns; Tosoh
Catalog No. 50-851-127