Learn More
Medchemexpress LLC Endo-1,4-β-xylanase | 9025-57-4 | ~22 kDa | 10 G
SDP
Supplier:  Medchemexpress LLC HYP301710G
Encompass_Preferred
Endo-1,4-β-xylanase (Xylanase) is an arabinoxylan (AX) degrading enzyme and a glycoside hydrolase. It is often utilized in biochemical studies due to its ability to cleave the β-xylosidic bond between two d-xylopyranosyl residues linked in β-(1,4).
  • Degrades arabinoxylan
  • Functions as a glycoside hydrolase
  • Applied in biochemical research
  • Facilitates the cleavage of β-xylosidic bonds
  • Sourced from Penicillium
  • Appears as a yellow to brown solid
Catalog No. 50-003-96600