Learn More
Medchemexpress LLC endo-BCN-PEG2-C2-NHS ester | 2243565-12-8 | 95.0% | 450.48 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140067
Encompass_Preferred
endo-BCN-PEG2-C2-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs consist of two different ligands connected by a linker, enabling the exploitation of the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-50164