Learn More
Medchemexpress LLC Endo-BCN-PEG2-NH2 | 1263166-93-3 | 97.0% | 324.42 | C17H28N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0727525MG
Encompass_Preferred
endo-BCN-PEG2-NH2 is a PEG-based PROTAC linker that contains a bicyclo[6.1.0]non-4-yne (BCN) moiety, enabling strain-promoted alkyne-azide cycloaddition (SPAAC) for copper-free click conjugation. It is intended for use in targeted protein degradation research and bioconjugation workflows.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) for copper-free click conjugation.
  • Contains a PEG2 spacer to improve solubility and provide flexibility.
  • Primary amine functional group allows straightforward coupling chemistry.
  • Suitable for small-scale synthesis and bioconjugation applications.
  • Available in low-milligram quantities for research use.
Catalog No. 50-002-09810