Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC endo-BCN-PEG24-NHS ester | C66H118N2O30 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1503115MG

Encompass_Preferred

endo-BCN-PEG24-NHS ester is a PROTAC linker composed of a benzocyclonorbornene, a twenty-tetramer of polyethylene glycol (PEG), and an N-hydroxysuccinimide ester. It is intended for research use only.

  • Utilized as linker for synthesis of PROTAC molecule.
  • Molecular weight is 1419.64.
  • Appearance is an oil, white to off-white in color.
  • Purity is ≥95.0%.
  • Ships at room temperature in the continental US.
  • Pure form storage: -20°C for 3 years.
  • In solvent storage: -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-003-75338


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.