Learn More
Medchemexpress LLC Endo-BCN-PEG24-NHS ester | >95.0% | 1419.64 | C66H118N2O30 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15031110MG
Encompass_Preferred
endo-BCN-PEG24-NHS ester is a heterobifunctional PEG24 linker bearing a benzocyclonorbornene (BCN) group and an N-hydroxysuccinimide (NHS) ester. It is designed for bioconjugation workflows, enabling strain-promoted alkyne-azide cycloaddition (SPAAC) and amine coupling for synthesis of conjugates such as PROTACs and labeled biomolecules. Intended for research use with supporting documentation.
Catalog No. 50-002-86223