Learn More
Medchemexpress LLC Endo-BCN-PEG4-PFP ester | 2904607-26-5 | 95.0% | 607.56 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140073
Encompass_Preferred
endo-BCN-PEG4-PFP ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It acts as a click chemistry reagent, featuring a BCN group that enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. This mechanism supports the development of PROTACs, which degrade target proteins via the ubiquitin-proteasome system. This product is for research use only.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains a BCN group for SPAAC with azide groups
  • Used in PROTAC synthesis
  • Enables selective degradation of target proteins
  • Functions through the intracellular ubiquitin-proteasome system
  • Intended for research use only
Catalog No. 50-003-51700