Learn More
Medchemexpress LLC Endo-BCN-PEG8-NHS ester | 1608140-48-2 | 95.0% | C34H54N2O14 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14007010MG
Encompass_Preferred
endo-BCN-PEG8-NHS ester is a bifunctional PEG8 linker that combines an endo bicyclononyne (BCN) strained alkyne with an N-hydroxysuccinimide (NHS) ester. It enables copper-free bioorthogonal conjugation via strain-promoted alkyne-azide cycloaddition (SPAAC) and efficient NHS-mediated amide bond formation for labeling or linking amine-containing molecules. Supplied as a high-purity liquid, it is intended for linker synthesis, bioconjugation, and molecular labeling workflows.
  • Facilitates copper-free SPAAC bioorthogonal conjugation.
  • Provides an active NHS ester for rapid amine coupling.
  • Contains a BCN strained alkyne for efficient click reactions.
  • Includes a PEG8 spacer to improve solubility and reduce steric hindrance.
  • Offered as a high-purity reagent suitable for synthesis and labeling.
  • Stable when stored sealed at recommended temperatures.
Catalog No. 50-002-60400