Learn More
LUNA NANOTECH INC Endo-Free Plasmid Maxiprep Kit, 30 Preps
Supplier:  LUNA NANOTECH INC NKE11330
Encompass_Preferred
PuroSPIN™ Endo-Free Plasmid MAXIprep Kit, 30 Preps
The PuroSPIN™ Endo-Free Plasmid MAXIprep Kit is designed to provide a simple and easy-to-use method for large-scale extraction and purification of endotoxin-free plasmid DNA.
This purification technology is based on plasmid binding to silica spin-columns, which allows for efficient removal of genomic DNA, protein, salt, and other contaminants. Following the lysis steps, endotoxins are removed using the Endotoxin Removal Buffer ERB.
The PuroSPIN™ Endo-Free Plasmid MAXIprep Kit can extraxt up to 500 µg of plasmid DNA with endotoxin levels of <0.1 EU/µg. Silica spin columns provide a faster, simpler, and safer alternative to traditional nucleic acid purification techniques such as phenol-chloroform extraction.
The PuroSPIN™ buffer formulations produce highly purified plasmid DNA for downstream applications, such as transfection, enzymatic digestion, or
Catalog No. 50-256-6925