Learn More
New England Biolabs, Inc. Endo H – 50000 units
SDP
Supplier:  New England Biolabs, Inc. P0702L
Encompass
Endoglycosidase H is a recombinant glycosidase which cleaves within the chitobiose core of high mannose and some hybrid oligosaccharides from N-linked glycoproteins.
Catalog No. 50-811-825